siRNA Details
Gene Symbol: S100a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108557.2 | NP_001102027.1 | 2 | 177 | ||
XM_006232717.2 | XP_006232779.1 | 2 | 251 | |||
XM_006232718.1 | XP_006232780.1 | 2 | 168 | |||
GenBank mRNA | BC158598.1 | - | 2 | 177 | 7127572 | |
FQ209774.1 | - | 2 | 179 | |||
FQ214291.1 | - | 2 | 160 | |||
FQ214430.1 | - | 2 | 173 | |||
FQ229313.1 | - | 2 | 185 |
