siRNA Details
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025018.1 | NP_001020189.1 | 7 | 852 | ||
XM_006233120.2 | XP_006233182.1 | 7 | 796 | |||
XM_006233121.2 | XP_006233183.1 | 6 | 736 | |||
XM_006233122.2 | XP_006233184.1 | 6 | 837 | |||
GenBank mRNA | BC091251.1 | - | 7 | 852 | 7372838 | |
FQ218349.1 | - | 7 | 864 | |||
FQ228782.1 | - | 7 | 949 |
