siRNA Details
Gene Symbol: Sypl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108563.1 | NP_001102033.1 | 3, 4 | 434 | ||
GenBank mRNA | AB158471.1 | - | 3, 4 | 252 |

More Information
Other Species Targeted

Gene Ontology Categories:
