siRNA Details
Gene Symbol: Ank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761478.1 | XP_008759700.1 | 4, 5 | 4789 | ||
GenBank mRNA | U65916.1 | - | 3, 4 | 286 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761478.1 | XP_008759700.1 | 4, 5 | 4789 | ||
GenBank mRNA | U65916.1 | - | 3, 4 | 286 |