siRNA Details
Gene Symbol: Unc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199407.1 | NP_955439.1 | 14 | 2474 | ||
XM_006233402.2 | XP_006233464.1 | 15 | 2479 | |||
GenBank mRNA | AB118026.1 | - | 14 | 2474 |
