siRNA Details
Gene Symbol: Setx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233886.2 | XP_006233948.1 | 10 | 2059 | ||
XM_006233887.2 | XP_006233949.1 | 11 | 2062 | |||
XM_006233888.2 | XP_006233950.1 | 10 | 2058 | |||
XM_008761681.1 | XP_008759903.1 | 10 | 2173 | |||
XM_008761682.1 | XP_008759904.1 | 7 | 1427 |
