siRNA Details
Gene Symbol: Lypd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134580.1 | NP_001128052.1 | 8, 9 | 742 | ||
XM_008761762.1 | XP_008759984.1 | 5, 6 | 2588 | |||
XM_008761763.1 | XP_008759985.1 | 6, 7 | 3238 | |||
XM_008761764.1 | XP_008759986.1 | 5, 6 | 3054 | |||
XM_008761765.1 | XP_008759987.1 | 7, 8 | 3358 | |||
XM_008761766.1 | XP_008759988.1 | 4, 5 | 2528 | |||
XM_008761767.1 | XP_008759989.1 | 4, 5 | 2994 | |||
XM_008761768.1 | XP_008759990.1 | 4, 5 | 2038 | |||
XM_008761769.1 | XP_008759991.1 | 5, 6 | 2494 |
