siRNA Details
Gene Symbol: Lypd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134580.1 | NP_001128052.1 | 7 | 565 | ||
XM_008761762.1 | XP_008759984.1 | 4 | 2411 | |||
XM_008761763.1 | XP_008759985.1 | 5 | 3061 | |||
XM_008761764.1 | XP_008759986.1 | 4 | 2877 | |||
XM_008761765.1 | XP_008759987.1 | 6 | 3181 | |||
XM_008761766.1 | XP_008759988.1 | 3 | 2351 | |||
XM_008761767.1 | XP_008759989.1 | 3 | 2817 | |||
XM_008761768.1 | XP_008759990.1 | 3 | 1861 | |||
XM_008761769.1 | XP_008759991.1 | 4 | 2317 |
