siRNA Details
Gene Symbol: Mmadhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004280.1 | NP_001004280.1 | 8 | 898 | ||
XM_006234158.2 | XP_006234220.1 | 8 | 1017 | |||
GenBank mRNA | BC078953.1 | - | 8 | 898 | 7110154 | |
FQ212704.1 | - | 8 | 893 | |||
FQ213711.1 | - | 8 | 876 | |||
FQ216239.1 | - | 8 | 893 | |||
FQ219117.1 | - | 8 | 894 | |||
FQ228704.1 | - | 8 | 894 | |||
FQ229971.1 | - | 8 | 863 |
