siRNA Details
Gene Symbol: Mmadhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004280.1 | NP_001004280.1 | 8 | 819 | ||
XM_006234158.2 | XP_006234220.1 | 8 | 938 | |||
GenBank mRNA | BC078953.1 | - | 8 | 819 | 7110154 | |
FQ212704.1 | - | 8 | 814 | |||
FQ213711.1 | - | 8 | 797 | |||
FQ216239.1 | - | 8 | 814 | |||
FQ219117.1 | - | 8 | 815 | |||
FQ228704.1 | - | 8 | 815 | |||
FQ229971.1 | - | 8 | 784 |
