siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025421.1 | NP_001020592.1 | 6, 7 | 705 | ||
XM_006234513.2 | XP_006234575.1 | 8, 9 | 1636 | |||
XM_006234514.1 | XP_006234576.1 | 7, 8 | 846 | |||
XM_006234516.1 | XP_006234578.1 | 7, 8 | 846 | |||
XM_006234518.1 | XP_006234580.1 | 7, 8 | 846 | |||
XM_006234522.1 | XP_006234584.1 | 7, 8 | 768 | |||
XM_006234523.1 | XP_006234585.1 | 8, 9 | 904 | |||
XM_006234524.2 | XP_006234586.1 | 8, 9 | 795 | |||
XM_006234526.2 | XP_006234588.1 | 8, 9 | 1019 | |||
XM_006234527.2 | XP_006234589.1 | 8, 9 | 1019 | |||
XM_008762008.1 | XP_008760230.1 | 9, 10 | 1838 | |||
XM_008762009.1 | XP_008760231.1 | 9, 10 | 921 | |||
XM_008762010.1 | XP_008760232.1 | 8, 9 | 1685 | |||
XM_008762011.1 | XP_008760233.1 | 7, 8 | 715 | |||
GenBank mRNA | BC098012.1 | - | 6, 7 | 705 | 7454358 | |
FQ222063.1 | - | 7, 8 | 847 |
