siRNA Details
Gene Symbol: Prr5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080150.1 | NP_001073619.1 | 9 | 1109 | ||
XM_006234647.2 | XP_006234709.1 | 9 | 2075 | |||
XM_006234648.2 | XP_006234710.1 | 9 | 1376 | |||
GenBank mRNA | AY724495.1 | - | 4 | 383 | ||
BC129104.1 | - | 9 | 1109 | 7100178 |
