siRNA Details
Gene Symbol: Prr5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080150.1 | NP_001073619.1 | 4, 5 | 650 | ||
XM_006234647.2 | XP_006234709.1 | 4, 5 | 1616 | |||
XM_006234648.2 | XP_006234710.1 | 4, 5 | 917 | |||
GenBank mRNA | BC129104.1 | - | 4, 5 | 650 | 7100178 |
