siRNA Details
Gene Symbol: Cstf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077672.1 | NP_001071140.1 | 13, 14 | 1267 | ||
XM_008762067.1 | XP_008760289.1 | 14, 15 | 1286 | |||
GenBank mRNA | BC127520.1 | - | 13, 14 | 1267 | 7110287 | |
FQ223256.1 | - | 8, 9 | 763 |
