siRNA Details
Gene Symbol: Ccdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108587.1 | NP_001102057.1 | 2 | 585 | ||
GenBank mRNA | FQ225525.1 | - | 2 | 451 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108587.1 | NP_001102057.1 | 2 | 585 | ||
GenBank mRNA | FQ225525.1 | - | 2 | 451 |