siRNA Details
Gene Symbol: Ccdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108587.1 | NP_001102057.1 | 5 | 1766 | ||
GenBank mRNA | FQ225525.1 | - | 6 | 1137 |
Gene Symbol: LOC102551066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234694.1 | XP_006234756.1 | 2 | 420 |
