siRNA Details
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234794.2 | XP_006234856.1 | 2 | 533 | ||
XM_006234796.2 | XP_006234858.1 | 2 | 2334 | |||
XM_008762115.1 | XP_008760337.1 | 2 | 203 | |||
XM_008762116.1 | XP_008760338.1 | 2 | 203 | |||
XM_008762117.1 | XP_008760339.1 | 2 | 203 |

More Information
Other Species Targeted

Gene Ontology Categories:
