siRNA Details
Gene Symbol: Snrpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108592.2 | NP_001102062.1 | 3 | 197 | ||
XM_006235116.2 | XP_006235178.1 | 3 | 358 | |||
GenBank mRNA | BC168760.1 | - | 3 | 197 | 6888886 | |
FQ213679.1 | - | 3 | 229 |
