siRNA Details
Gene Symbol: Naa20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108595.1 | NP_001102065.1 | 2, 3 | 145 | ||
XM_006235155.2 | XP_006235217.1 | 2, 3 | 144 | |||
GenBank mRNA | BC167020.1 | - | 2, 3 | 105 | 9025632 | |
FQ234868.1 | - | 2, 3 | 100 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001141965, NM_026425, BC116374, AK005143, BC009157, and others.
Gene Ontology Categories:
