siRNA Details
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108598.2 | NP_001102068.2 | 2 | 246 | ||
NM_001244784.1 | NP_001231713.1 | 2 | 246 | |||
GenBank mRNA | BC168211.1 | - | 2 | 75 | 7135378 | |
CB576376.1 | - | 2 | 123 | |||
CB585518.1 | - | 2 | 246 | |||
CB736780.1 | - | 2 | 325 |
