siRNA Details
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108598.2 | NP_001102068.2 | 5, 6 | 464 | ||
GenBank mRNA | CB576376.1 | - | 5, 6 | 341 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108598.2 | NP_001102068.2 | 5, 6 | 464 | ||
GenBank mRNA | CB576376.1 | - | 5, 6 | 341 |