siRNA Details
Gene Symbol: Ift52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177685.1 | NP_001171156.1 | 5, 6 | 466 | ||
GenBank mRNA | BC162007.1 | - | 5, 6 | 466 | 9090205 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ift52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177685.1 | NP_001171156.1 | 5, 6 | 466 | ||
GenBank mRNA | BC162007.1 | - | 5, 6 | 466 | 9090205 |