siRNA Details
Gene Symbol: Rbpjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108604.1 | NP_001102074.1 | 12 | 1559 | ||
XM_006235598.2 | XP_006235660.1 | 11 | 1501 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbpjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108604.1 | NP_001102074.1 | 12 | 1559 | ||
XM_006235598.2 | XP_006235660.1 | 11 | 1501 |