siRNA Details
Gene Symbol: Myt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108615.1 | NP_001102085.1 | 15 | 3419 | ||
XM_006235782.1 | XP_006235844.1 | 21 | 3236 | |||
XM_006235783.2 | XP_006235845.1 | 21 | 3470 | |||
XM_006235784.1 | XP_006235846.1 | 20 | 3167 | |||
XM_006235785.2 | XP_006235847.1 | 21 | 3187 | |||
XM_006235787.2 | XP_006235849.1 | 18 | 3057 |
