siRNA Details
Gene Symbol: Agap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108616.1 | NP_001102086.1 | 10 | 2169 | ||
XM_006235938.2 | XP_006236000.1 | 13 | 2279 | |||
XM_006235941.2 | XP_006236003.1 | 13 | 1968 | |||
XM_008762650.1 | XP_008760872.1 | 13 | 1445 |
