siRNA Details
Gene Symbol: Agap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108616.1 | NP_001102086.1 | 13 | 2673 | ||
XM_006235938.2 | XP_006236000.1 | 16 | 2783 | |||
XM_006235939.2 | XP_006236001.1 | 15 | 2644 | |||
XM_006235940.2 | XP_006236002.1 | 14 | 2478 | |||
XM_006235941.2 | XP_006236003.1 | 16 | 2472 | |||
XM_006235942.2 | XP_006236004.1 | 14 | 2168 | |||
XM_008762650.1 | XP_008760872.1 | 16 | 1949 | |||
GenBank mRNA | BC158716.1 | - | 1 | 54 | ||
KF447874.1 | - | 14 | 1874 |
