siRNA Details
Gene Symbol: Impdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108619.1 | NP_001102089.1 | 7, 8 | 759 | ||
XM_006236208.1 | XP_006236270.1 | 7, 8 | 981 | |||
XM_006236210.1 | XP_006236272.1 | 7, 8 | 981 | |||
XM_008762791.1 | XP_008761013.1 | 7, 8 | 984 | |||
XM_008762792.1 | XP_008761014.1 | 6, 7 | 909 | |||
XM_008762793.1 | XP_008761015.1 | 7, 8 | 984 | |||
XM_008762794.1 | XP_008761016.1 | 6, 7 | 909 |
