siRNA Details
Gene Symbol: Tcaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749739.3 | XP_003749787.1 | 6 | 2305 | ||
XM_008762928.1 | XP_008761150.1 | 6 | 2634 | |||
XM_342675.6 | XP_342676.4 | 6 | 2634 | |||
GenBank mRNA | FQ211472.1 | - | 6 | 2272 |
