siRNA Details
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108627.2 | NP_001102097.1 | 7 | 927 | ||
XM_006236409.1 | XP_006236471.1 | 6 | 835 | |||
XM_006236410.2 | XP_006236472.1 | 6 | 837 | |||
XM_006236411.1 | XP_006236473.1 | 6 | 835 | |||
GenBank mRNA | BC161932.1 | - | 7 | 927 | 9021933 |
