siRNA Details
Gene Symbol: Rad18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077673.1 | NP_001071141.1 | 7 | 890 | ||
XM_006237036.1 | XP_006237098.1 | 7 | 919 | |||
XM_006237037.1 | XP_006237099.1 | 7 | 919 | |||
GenBank mRNA | BC127510.1 | - | 7 | 890 | 8366167 | |
EF395817.1 | - | 860 |
