siRNA Details
Gene Symbol: Iffo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108647.1 | NP_001102117.1 | 1 | 19 | ||
XM_006237372.2 | XP_006237434.1 | 1 | 50 | |||
XM_006237373.2 | XP_006237435.1 | 1 | 50 | |||
XM_006237374.2 | XP_006237436.1 | 1 | 51 | |||
XM_006237375.2 | XP_006237437.1 | 1 | 50 | |||
XM_006237376.2 | XP_006237438.1 | 1 | 50 | |||
XM_008763292.1 | XP_008761514.1 | 1 | 51 |
