siRNA Details
Gene Symbol: Itfg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108648.1 | NP_001102118.1 | 11 | 1265 | ||
XM_006237409.2 | XP_006237471.1 | 11 | 1654 | |||
XM_006237410.2 | XP_006237472.1 | 10 | 1601 | |||
GenBank mRNA | BC158714.1 | - | 10 | 1120 | 9088606 |
