siRNA Details
Gene Symbol: Arhgdib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009600.1 | NP_001009600.1 | 6 | 643 | ||
XM_006237561.2 | XP_006237623.1 | 6 | 667 | |||
XM_006237562.2 | XP_006237624.1 | 6 | 798 | |||
XM_006237563.2 | XP_006237625.1 | 7 | 729 | |||
XM_006237564.2 | XP_006237626.1 | 6 | 758 | |||
XM_006237565.2 | XP_006237627.1 | 6 | 943 | |||
GenBank mRNA | BC088209.1 | - | 6 | 643 | 7382227 | |
FQ225418.1 | - | 6 | 617 | |||
FQ233135.1 | - | 6 | 624 | |||
FQ234136.1 | - | 6 | 638 |
