siRNA Details
Gene Symbol: Golt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113783.1 | NP_001107255.1 | 2, 3 | 197 | ||
GenBank mRNA | BC158751.1 | - | 2, 3 | 197 | 9102153 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Golt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113783.1 | NP_001107255.1 | 2, 3 | 197 | ||
GenBank mRNA | BC158751.1 | - | 2, 3 | 197 | 9102153 |