siRNA Details
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108657.1 | NP_001102127.1 | 5 | 973 | ||
XM_006237901.2 | XP_006237963.1 | 6 | 1106 | |||
XM_006237903.2 | XP_006237965.1 | 5 | 878 | |||
XM_006237904.2 | XP_006237966.1 | 5 | 936 | |||
XM_008763548.1 | XP_008761770.1 | 3 | 2384 | |||
XM_008763549.1 | XP_008761771.1 | 5 | 6269 | |||
XM_008763550.1 | XP_008761772.1 | 6 | 802 |
