siRNA Details
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108657.1 | NP_001102127.1 | 6 | 1068 | ||
XM_006237901.2 | XP_006237963.1 | 7 | 1201 | |||
XM_006237903.2 | XP_006237965.1 | 6 | 973 | |||
XM_006237904.2 | XP_006237966.1 | 6 | 1031 | |||
XM_008763548.1 | XP_008761770.1 | 4 | 2479 | |||
XM_008763549.1 | XP_008761771.1 | 6 | 6364 | |||
XM_008763550.1 | XP_008761772.1 | 7 | 897 |
