siRNA Details
Gene Symbol: Tmem246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014190.1 | NP_001014212.1 | 1 | 663 | ||
XM_006238158.2 | XP_006238220.1 | 2 | 4015 | |||
XM_006238159.2 | XP_006238221.1 | 2 | 1136 | |||
GenBank mRNA | BC089972.1 | - | 1 | 663 | 7315559 |
