siRNA Details
Gene Symbol: Smc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108666.3 | NP_001102136.2 | 25 | 3613 | ||
XM_006238165.2 | XP_006238227.1 | 24 | 3557 | |||
GenBank mRNA | CV074555.1 | - | 5 | 530 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008017, BC094380, AJ534939, AK145341, BC071232, and others.
Gene Ontology Categories:
