siRNA Details
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108667.1 | NP_001102137.1 | 5 | 745 | ||
XM_006238166.2 | XP_006238228.1 | 7 | 852 | |||
XM_006238168.2 | XP_006238230.1 | 6 | 779 | |||
XM_006238169.1 | XP_006238231.1 | 5 | 662 | |||
XM_008763712.1 | XP_008761934.1 | 6 | 872 | |||
XM_008763713.1 | XP_008761935.1 | 7 | 846 | |||
XM_008763714.1 | XP_008761936.1 | 6 | 780 | |||
XM_008763715.1 | XP_008761937.1 | 6 | 779 |
