siRNA Details
Gene Symbol: Akna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108668.1 | NP_001102138.1 | 3 | 1395 | ||
XM_006238255.2 | XP_006238317.1 | 2 | 1261 | |||
XM_006238256.2 | XP_006238318.1 | 3 | 1740 | |||
XM_006238257.2 | XP_006238319.1 | 3 | 1306 | |||
XM_008763780.1 | XP_008762002.1 | 3 | 1308 | |||
XM_008763781.1 | XP_008762003.1 | 4 | 3728 | |||
XM_008763782.1 | XP_008762004.1 | 4 | 3189 | |||
XM_008763783.1 | XP_008762005.1 | 3 | 2449 | |||
XM_008763784.1 | XP_008762006.1 | 3 | 3058 |
