siRNA Details
Gene Symbol: Hacd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108669.1 | NP_001102139.1 | 3 | 264 | ||
XM_006238376.2 | XP_006238438.1 | 3 | 298 | |||
XM_008763794.1 | XP_008762016.1 | 3 | 308 | |||
XM_008763795.1 | XP_008762017.1 | 3 | 307 | |||
GenBank mRNA | FQ234476.1 | - | 3 | 260 |
