siRNA Details
Gene Symbol: Efcab7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191866.1 | NP_001178795.1 | 5 | 657 | ||
XM_008763885.1 | XP_008762107.1 | 5 | 707 | |||
XM_008763886.1 | XP_008762108.1 | 5 | 645 |
