siRNA Details
Gene Symbol: Pomgnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007747.1 | NP_001007748.1 | 6 | 670 | ||
XM_006238683.2 | XP_006238745.1 | 5 | 777 | |||
GenBank mRNA | BC083641.1 | - | 6 | 670 | 7134894 |
