siRNA Details
Gene Symbol: Zmym6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108681.2 | NP_001102151.1 | 7 | 1036 | ||
XM_006238892.2 | XP_006238954.1 | 6 | 1867 | |||
XM_006238894.2 | XP_006238956.1 | 7 | 914 | |||
GenBank mRNA | FQ221018.1 | - | 6 | 1402 |
