siRNA Details
Gene Symbol: Pum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108684.1 | NP_001102154.1 | 6 | 959 | ||
XM_006239029.1 | XP_006239091.1 | 6 | 986 | |||
XM_006239030.2 | XP_006239092.1 | 6 | 984 | |||
XM_006239031.1 | XP_006239093.1 | 6 | 986 | |||
XM_006239032.1 | XP_006239094.1 | 6 | 986 | |||
XM_006239033.1 | XP_006239095.1 | 6 | 986 | |||
XM_006239034.1 | XP_006239096.1 | 6 | 986 | |||
XM_006239035.1 | XP_006239097.1 | 6 | 986 | |||
XM_006239036.1 | XP_006239098.1 | 6 | 986 | |||
XM_006239037.2 | XP_006239099.1 | 4 | 699 | |||
XM_006239038.2 | XP_006239100.1 | 4 | 699 | |||
XM_008764150.1 | XP_008762372.1 | 6 | 960 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030722, NM_001159603, NM_001159604, NM_001159605, BC048174, and others.
Gene Ontology Categories:
