siRNA Details
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014194.1 | NP_001014216.1 | 15 | 2052 | ||
GenBank mRNA | BC089875.1 | - | 15 | 2052 | 7325475 |

More Information
Other Species Targeted

Gene Ontology Categories:
