siRNA Details
Gene Symbol: Srsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039035.2 | NP_001034124.2 | 1, 2 | 122 | ||
XM_006239642.1 | XP_006239704.1 | 1, 2 | 162 | |||
XM_006239643.1 | XP_006239705.1 | 1, 2 | 162 | |||
XM_006239644.1 | XP_006239706.1 | 1, 2 | 162 | |||
XM_006239645.1 | XP_006239707.1 | 1, 2 | 162 | |||
XM_006239646.1 | XP_006239708.1 | 1, 2 | 162 | |||
GenBank mRNA | BC099175.1 | - | 1, 2 | 112 | 7382701 | |
FM084661.1 | - | 1, 2 | 122 | |||
FM085512.1 | - | 1, 2 | 116 | |||
FQ219847.1 | - | 1, 2 | 128 | |||
FQ229134.1 | - | 1, 2 | 129 |
