siRNA Details
Gene Symbol: Spast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108702.2 | NP_001102172.2 | 7, 8 | 1082 | ||
XM_008764486.1 | XP_008762708.1 | 8, 9 | 1458 | |||
XM_008764487.1 | XP_008762709.1 | 8, 9 | 1455 | |||
XM_008764488.1 | XP_008762710.1 | 7, 8 | 1363 | |||
GenBank mRNA | BC166846.1 | - | 7, 8 | 1082 | 9030077 |
