siRNA Details
Gene Symbol: Wdr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037791.2 | NP_001032880.2 | 7 | 837 | ||
XM_006239770.2 | XP_006239832.1 | 7 | 845 | |||
GenBank mRNA | BC169117.1 | - | 7 | 818 |
