siRNA Details
Gene Symbol: Dtnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012191.1 | NP_001012191.1 | 9 | 1187 | ||
XM_006239816.2 | XP_006239878.1 | 9 | 1126 | |||
XM_006239817.2 | XP_006239879.1 | 9 | 1125 | |||
XM_006239818.2 | XP_006239880.1 | 9 | 1125 | |||
XM_006239819.2 | XP_006239881.1 | 9 | 1126 | |||
XM_006239822.2 | XP_006239884.1 | 9 | 1125 | |||
GenBank mRNA | BC081889.1 | - | 9 | 1187 | 7109493 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001162465, NM_007886, Y15742, BC030924, AK043436, and others.
Gene Ontology Categories:
